Learn More
RETSCH Receptacles for FGM 200 Knife Mills
Product Code. 10340912 Shop All Retsch Products
Change view
Click to view available options
Material:
Glass
Stainless Steel
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material unitSize
10340912 Glass Each
10085271 Stainless Steel Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10340912 Supplier RETSCH Supplier No. 03.045.0046
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Vessel for high-speed knife mills
Available in glass and stainless steel.
Specifications
Product Type Receptacle
For Use With (Equipment) Grindomix GM 200 knife mill
Material Glass