missing translation for 'onlineSavingsMsg'
Learn More

Retinoblastoma (Rb) (Phospho-specific Serine) Ab-7, Mouse Monoclonal Antibody,Lab Vision™ Epredia™

Provide accurate, reproducible results with the Epredia™ Retinoblastoma (Rb) (Phospho-specific Serine) Ab-7, Mouse Monoclonal Antibody.

£122.40 - £766.00

Specifications

Antigen Retinoblastoma (Rb) (Phospho-specific Serine) Ab-7
Clone 51B7
Applications Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 4
Product Code Brand Quantity Concentration Price Quantity & Availability  
Product Code Brand Quantity Concentration Price Quantity & Availability  
12650397
View Documents
Epredia
MS-1883-P0
100 μL 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12680397
View Documents
Epredia
MS-1883-P1
500 μL 200μg/mL
ONLINE EXCLUSIVE PRICE
£513.00
£483.40 / 500µL
Save £29.60
5% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
12670397
missing translation for 'viewDocuments'
Epredia
MS-1883-P1ABX
100 μL 1mg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A Ver productos alternativos
missing translation for 'toSaveUpto' 
»
12690397
missing translation for 'viewDocuments'
Epredia
MS-1883-PABX
200 μL 1mg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A Ver productos alternativos
missing translation for 'toSaveUpto' 
»
Descripción

Descripción

Rb is a tumor suppressor nuclear phosphoprotein capable of binding to DNA. It is phosphorylated on serine and threonine, but not on tyrosine residues. It forms a complex with SV40 large T antigen, adenovirus E1A, and human papilloma virus-16 E. Rb protein may act by regulating transcription and loss of its function leads to uncontrolled cell growth. Aberrations in the RB gene have been implicated in cancers of breast, colon, prostate, kidney, nasopharynx, and leukemia. Phosphorylation of Rb at S608 depends on CDK4.

Host Species: Mouse

Clone: 51B7

Isotype: IgG1

Species Reactivity: Human and Mouse. Others not known.

Molecular Weight: 105kDa

Positive Control: Ls174T cells

Cellular Localization: Nuclear Ab-7 reacts with human and mouse protein phosphorylated on S608. It does not react with nonphosphorylate Rb or Rb phosphorylated at other sites.

Recommended for:

  • Western Blotting

Note:

USA: RUO; Int'l: RUO

Especificaciones

Especificaciones

Retinoblastoma (Rb) (Phospho-specific Serine) Ab-7
Western Blot
Unconjugated
Mouse
Cancer and Tumor Biology
IgG1
51B7
Monoclonal
Concentrated
RUO
Human, Mouse
Primary
Videos
SDS
Documentos

Documentos

Certificados
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado