Learn More
Retinoblastoma (Rb) (Phospho-specific Serine) Ab-7, Mouse Monoclonal Antibody,Lab Vision™ Epredia™
Shop All Epredia ProductsDescription
Host Species: Mouse
Clone: 51B7
Isotype: IgG1
Species Reactivity: Human and Mouse. Others not known.
Molecular Weight: 105kDa
Positive Control: Ls174T cells
Cellular Localization: Nuclear Ab-7 reacts with human and mouse protein phosphorylated on S608. It does not react with nonphosphorylate Rb or Rb phosphorylated at other sites.
Recommended for:
- Western Blotting
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Retinoblastoma (Rb) (Phospho-specific Serine) Ab-7 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 51B7 |
| Concentration | 200μg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Quantity | 500 μL |
| Regulatory Status | RUO |
| Research Discipline | Cancer and Tumor Biology |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.