Learn More
Honeywell™ Retaining Covers for Prefilter Pads
Retaining Covers for Prefilter Pads are used in conjunction with Honeywell™ cartridges on twin-filter respirators
Brand: Honeywell™ 1001606
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- Product Type: Retaining Cover
- Color: White
- Material: Plastic
- Requires: Prefilter Pad
Specifications
Pre-Assembled Cartridges on Freedom Series Respirators | |
Requires Honeywell™ Prefilter Pads [11847171 (P1) / 12682743 (P2)] | |
Honeywell™ Freedom Half-Mask Air-Purifying Respirators |
White | |
Retainer Cover for Prefilter Pads | |
RP78 |