Learn More
Agilent BioTek Retainer Clip for Synergy and FLx800
Product Code. 12386453 Shop All Agilent BioTek Products
Click to view available options
For Use With (Equipment):
Synergy, FLx800
Unit Size:
Pack of 5
Product Code. 12386453
Brand: Agilent BioTek 7082075
Retainer Clip for Synergy and FLx800 holds filters and plugs in place in the filter wheels.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Retainer Clip
For Use With (Equipment) Synergy, FLx800
For Use With (Application) Holds Filters and Plugs in Filter Wheels