Learn More
Restek™ Tube and Pipe Fittings for GC Equipment
Ideal for installing new equipment, modifying existing instrumentation, or replacing worn connections.
Brand: Restek™ 21810
Additional Details : Weight : 0.25000kg
Description
- Available in brass and stainless steel
- Siltek/Sulfinert treated or Silcosteel-CR treated fittings
Note
Nuts and ferrules are not treated unless requested. Nuts and ferrules normally are not in contact with sample pathway, and thus do not require coating.
Specifications
GC Equipment | |
0.125 in. | |
20/Pk. |
Brass | |
Nut and Ferrule Fitting Set | |
Nuts, Back Ferrules, and Front Ferrules |