Learn More
Restek™ Tube and Pipe Fittings for GC Equipment
Product Code. p-3207263
Encompass_Preferred
Change view
Click to view available options
Type:
Back Ferrule Fitting
Female Connector Fitting
Front Ferrule Fitting
Male Connector Fitting
Nut Fitting
Nut and Ferrule Set
Reducing Union Fitting
Tee Fitting
Diameter (English) Inside:
0.0625 to 0.125 in.
0.125 in.
0.125 to 0.0625 in.
0.25 in.
0.25 to 0.125 in.
Material:
Brass
Stainless Steel
Quantity:
1 Ea.
10/Pk.
2/Pk.
20/Pk.
40/Pk.
5/Pk.
Unit Size:
Pack of 10
Pack of 2
Pack of 20
Pack of 5
20 product options available for selection
Product selection table with 20 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Diameter (English) Inside Material Quantity unitSize
13169692 Nut Fitting 0.125 in. Brass 40/Pk. -
13199692 Front Ferrule Fitting 0.125 in. Brass 40/Pk. -
13109702 Front Ferrule Fitting 0.25 in. Brass 40/Pk. -
13139702 Back Ferrule Fitting 0.25 in. Brass 40/Pk. -
13129702 Back Ferrule Fitting 0.125 in. Brass 40/Pk. -
13169702 Nut and Ferrule Set 0.25 in. Brass 20/Pk. -
10584693 Nut and Ferrule Set 0.25 in. Brass 20/Pk. Pack of 20
10085281 Nut and Ferrule Set 0.125 in. Brass 20/Pk. Pack of 20
13159702 Nut and Ferrule Set 0.125 in. Brass 20/Pk. -
13139712 Reducing Union Fitting 0.125 to 0.0625 in. Brass 5/Pk. -
13179712 Tee Fitting 0.125 in. Brass 2/Pk. -
13189712 Tee Fitting 0.25 in. Brass 2/Pk. Pack of 2
13181312 Male Connector Fitting 0.125 in. Brass 10/Pk. Pack of 10
13101322 Male Connector Fitting 0.0625 to 0.125 in. Brass 5/Pk. -
10092644 Female Connector Fitting 0.25 to 0.125 in. Brass 5/Pk. Pack of 5
13151322 Female Connector Fitting 0.25 to 0.125 in. Brass 5/Pk. -
13151362 Nut and Ferrule Set 0.125 in. Stainless Steel 5/Pk. -
13161362 Nut and Ferrule Set 0.25 in. Stainless Steel 5/Pk. Pack of 5
13131372 Reducing Union Fitting 0.125 to 0.0625 in. Stainless Steel 1 Ea. -
13151372 Reducing Union Fitting 0.25 to 0.125 in. Stainless Steel 2/Pk. -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 20 options available.
20 options
Product Code. 13169692 Supplier Restek™ Supplier No. 21801
This item requires a price update prior to being ordered.
Request a Price for this item here.
Ideal for installing new equipment, modifying existing instrumentation, or replacing worn connections.
  • Available in brass and stainless steel
  • Siltek/Sulfinert treated or Silcosteel-CR treated fittings
Note:
Nuts and ferrules are not treated unless requested. Nuts and ferrules normally are not in contact with sample pathway, and thus do not require coating.
Specifications
Type Nut Fitting
Diameter (English) Inside 0.125 in.
For Use With (Equipment) GC Equipment
Material Brass
Quantity 40/Pk.