missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Restek™ Split Liners for Agilent™/Finnigan™ GCs: Cyclosplitter
For use with dirty samples
Brand: Restek™ 20708
Code : Z3
Additional Details : Weight : 1.00000kg
Description
- Incorporate a cylindrical glass screw in the sample pathway to mix sample and trap nonvolatile residues
- 4mm I.D.
- Perform multiple injections before cleaning is required
Specifications
IP CycloSplitter Inlet Liner | |
78.5 mm | |
6.3 mm | |
Borosilicate Glass |
25/Pk. | |
4.0 mm | |
Agilent GCs Equipped with Split/Splitless Inlets |