Learn More
Restek™ Resprep™-C18 and Resprep™-C8 SPE Disks
Extract analytes using high flow rates with significantly less clogging
Brand: Restek™ 24004
Code : Z3
Additional Details : Weight : 1.00000kg
Description
- Glass fiber disks embedded with C18 or C8 bonded silica
- Extract semivolatile organic compounds
- Deep-pore design reduces clogging and allows faster flow rates
- Meet requirements for EPA methods 525.1, 506, 550.1, 549.1
Specifications
EPA 525.1, 506, 550.1, 549.1 | |
SPE Disk | |
4.7 cm |
C18 | |
Glass Fiber |