Learn More
Restek™ Inlet Liners for Splitless Injection—Gooseneck Splitless
Useful for splitless mode of operation and slow injections, small (<2μL) sample volumes, and high-boiling point solvent systems
Brand:  Restek™ 20799-202.5
Code : Z3
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 12912849
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Compatible with Agilent™/Finnigan columns
  • Liners should be packed with wool or fused silica beads to trap nonvolatile residue and prevent column contamination when analyzing dirty samples
Specifications
Specifications
Glass Wool
5/Pk.
0.4 cm
Agilent GCs Equipped with Split/Splitless Inlets
IP Gooseneck Splitless Single Taper Inlet Liner; For trace samples greater than 2 μL
78.5 mm
0.65 cm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Ofertas especiales