missing translation for 'onlineSavingsMsg'
Learn More

Restek™ Inlet Liners for Splitless Injection—Gooseneck Splitless

Useful for splitless mode of operation and slow injections, small (<2μL) sample volumes, and high-boiling point solvent systems

Brand:  Restek™ 20799

Code : Z3

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 12902849

  • £112.53 / Pack of 5
Estimated Shipment: 17-05-2024
to see stock.
Add to basket

Description

Description

  • Compatible with Agilent™/Finnigan columns
  • Liners should be packed with wool or fused silica beads to trap nonvolatile residue and prevent column contamination when analyzing dirty samples

Specifications

Specifications

No FS Wool
5/Pk.
0.4 cm
Agilent GCs Equipped with Split/Splitless Inlets
IP Gooseneck Splitless Single Taper Inlet Liner
7.85 cm
0.65 cm
Borosilicate Glass
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers