Learn More
Restek™ Fittings, Parker (Treated): Tees
Ideal for installing new equipment, modifying existing instrumentation, or replacing worn connections.
Brand:  Restek™ 22868
Code : Z3
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 13137282
  • POA / Each
Estimated Shipment: 22-05-2024
to see stock.
This item requires a price update prior to being ordered.
Request a Price for this item here.
Description
Description
  • Available in Siltek/Sulfinert treated or Silcosteel-CR treated fittings
Specifications
Specifications
Tee
1/4 in.
Silcosteel-CR Coated