Learn More
Restek™ Fittings, Parker (Treated): Tees
Ideal for installing new equipment, modifying existing instrumentation, or replacing worn connections.
POA - £288.00
Product Code | Brand | Connections | Material | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Connections | Material | Price | Quantity & Availability | ||||
13174702
|
Restek™
22528 |
1/4 in. | Sulfinert-coated |
£177.60
Each |
Estimated Shipment: 24-04-2024
Log in to see stock. |
||||
13164702
|
Restek™
22527 |
1/8 in. | Sulfinert-coated |
£225.60
Each |
Estimated Shipment: 24-04-2024
Log in to see stock. |
||||
13154702
|
Restek™
22526 |
1/16 in. | Sulfinert-coated |
£288.00
Each |
Estimated Shipment: 24-04-2024
Log in to see stock. |
||||
13127282
|
Restek™
22867 |
1/8 in. | Silcosteel-CR Coated |
POA
Each |
N/A | ||||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
13137282
|
Restek™
22868 |
1/4 in. | Silcosteel-CR Coated |
POA
Each |
N/A | ||||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
13117282
|
Restek™
22866 |
1/16 in. | Silcosteel-CR Coated |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Available in Siltek/Sulfinert treated or Silcosteel-CR treated fittings
Specifications
Tee |