Learn More
Restek™ Corporation Vu2 Union™ Connector Kit Connectors
Vu2 Union connector combines the simplicity of a Press-Tight union with the strength of a metal union.
Brand:  Restek™ 21107
Additional Details : Weight : 0.99000kg
 View more versions of this product
Product Code. 10186683
  • £248.40 / Each
Estimated Shipment: 14-05-2024
to see stock.
Add to basket
Description
Description
  • Connect a guard column to an analytical column
  • Connect a column to a transfer line
  • Connect two columns in series
  • Repair a broken column
  • Fit both Restek cage designs
Specifications
Specifications
Connector
0.45 to 0.5 mm
Vu2 Union body, two knurled nuts, two Press-Tight unions, and four ferrules
Vu2 Union Connector Kit