Learn More
Restek™ Corporation Vu2 Union™ Connector Kit Connectors
Vu2 Union connector combines the simplicity of a Press-Tight union with the strength of a metal union.
Brand: Restek™ 21106
Code : Z3
Additional Details : Weight : 1.00000kg
Description
- Connect a guard column to an analytical column
- Connect a column to a transfer line
- Connect two columns in series
- Repair a broken column
- Fit both Restek cage designs
Specifications
Connector | |
0.28 to 0.32 mm | |
Vu2 Union body, two knurled nuts, two Press-Tight unions, and four ferrules | |
Vu2 Union Connector Kit |