Learn More
Invitrogen™ REST Monoclonal Antibody (K92059_15A3), Invitrogen™
Mouse Monoclonal Antibody
£380.00 - £570.00
Specifications
| Antigen | REST |
|---|---|
| Clone | K92059_15A3 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30199117
|
Invitrogen™
MA555715 |
50 μL |
£380.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30205246
|
Invitrogen™
MA556095 |
200 μL |
£570.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: PFKCDQCSYV ASNQHEVTRH ARQVHNGPKP LNCPHCDYKT ADRSNFKKHV ELHVNPRQFN CPVCDYAASK KCNLQYHFKS KHPTCPNKTM DVSKVKLKKT KKREADLPDN ITNEKTEIEQ TKIKGDVAGK KNEKSVKAEK RDVSKEKKPS NNVSVIQVTT RTRKSVTEVK EMDVHTGSNS EKFSKTKKSK RKLEVDSHSL HGPVNDEESS TKKKKKVESK SKNNSQEVPK GDSKVEENKK QNTCMKKSTK KKTLKNKSSK KSSKPPQKEP VEKGSAQMDP PQMGPAPTEA VQKGPVQVEP PPPMEHAQME
REST Monoclonal antibody specifically detects REST in Human samples. It is validated for Western BlotSpecifications
| REST | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| 2610008J04Rik; AA407358; Neural-restrictive silencer factor; neuron restrictive silencer factor; neuron-restrictive silencer factor; NRSF; RE1 silencing transcription factor; RE1-silencing transcription factor; RE1-silencing transcription factor variant E1a/E2/E3/E4c; RE1-silencing transcription factor variant E1a/E2/E3/E5; RE1-silencing transcription factor variant E1a/E2/E3/N3a/E4i; RE1-silencing transcription factor variant E1a/E2/E3/N3c/E4; RE1-silencing transcription factor variant E1a/E2/E4; RE1-silencing transcription factor variant E1a/E2/E5; RE1-silencing transcription factor variant E1a/E2a/E2k; RE1-silencing transcription factor variant E1a/E2d/E4g; RE1-silencing transcription factor variant E1a/E2e/E4h; RE1-silencing transcription factor variant E1a/E2f/E4e; RE1-silencing transcription factor variant E1a/E2k/E2i/E3/E4j; RE1-silencing transcription factor variant E1b/E2/E3/E5; RE1-silencing transcription factor variant E1b/E2/E3/N3b/E4i; RE1-silencing transcription factor variant E1b/E2/E3/N3c/E4; RE1-silencing transcription factor variant E1b/E2a/E2k; RE1-silencing transcription factor variant E1b/E2c/E2j/E3/E4; RE1-silencing transcription factor variant E1b/E2e/E4h; RE1-silencing transcription factor variant E1c/E2/E3/E5; RE1-silencing transcription factor variant E1c/E2a/E2k; RE1-silencing transcription factor variant E1c/E2g/E3/E4; repressor binding to the X2 box; Rest; REST4; WT6; X2 box repressor; XBR; zinc finger transcription factor REST protein | |
| Recombinant human REST (immunogen range is 331-640). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| REST |
| K92059_15A3 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q13127 | |
| 5978 | |
| IgG2a | |
| Protein A | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.