Learn More
Invitrogen™ REST Monoclonal Antibody (K92059_15A3), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA555715
Description
Sequence of this protein is as follows: PFKCDQCSYV ASNQHEVTRH ARQVHNGPKP LNCPHCDYKT ADRSNFKKHV ELHVNPRQFN CPVCDYAASK KCNLQYHFKS KHPTCPNKTM DVSKVKLKKT KKREADLPDN ITNEKTEIEQ TKIKGDVAGK KNEKSVKAEK RDVSKEKKPS NNVSVIQVTT RTRKSVTEVK EMDVHTGSNS EKFSKTKKSK RKLEVDSHSL HGPVNDEESS TKKKKKVESK SKNNSQEVPK GDSKVEENKK QNTCMKKSTK KKTLKNKSSK KSSKPPQKEP VEKGSAQMDP PQMGPAPTEA VQKGPVQVEP PPPMEHAQME
Transcriptional repressor which binds neuron-restrictive silencer element (NRSE) and represses neuronal gene transcription in non-neuronal cells. Restricts the expression of neuronal genes by associating with two distinct corepressors, mSin3 and CoREST, which in turn recruit histone deacetylase to the promoters of REST-regulated genes. Mediates repression by recruiting the BHC complex at RE1/NRSE sites which acts by deacetylating and demethylating specific sites on histones, thereby acting as a chromatin modifier.
Specifications
| REST | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Q13127 | |
| REST | |
| Recombinant human REST (immunogen range is 331-640). | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Western Blot | |
| K92059_15A3 | |
| Unconjugated | |
| REST | |
| 2610008J04Rik; AA407358; Neural-restrictive silencer factor; neuron restrictive silencer factor; neuron-restrictive silencer factor; NRSF; RE1 silencing transcription factor; RE1-silencing transcription factor; RE1-silencing transcription factor variant E1a/E2/E3/E4c; RE1-silencing transcription factor variant E1a/E2/E3/E5; RE1-silencing transcription factor variant E1a/E2/E3/N3a/E4i; RE1-silencing transcription factor variant E1a/E2/E3/N3c/E4; RE1-silencing transcription factor variant E1a/E2/E4; RE1-silencing transcription factor variant E1a/E2/E5; RE1-silencing transcription factor variant E1a/E2a/E2k; RE1-silencing transcription factor variant E1a/E2d/E4g; RE1-silencing transcription factor variant E1a/E2e/E4h; RE1-silencing transcription factor variant E1a/E2f/E4e; RE1-silencing transcription factor variant E1a/E2k/E2i/E3/E4j; RE1-silencing transcription factor variant E1b/E2/E3/E5; RE1-silencing transcription factor variant E1b/E2/E3/N3b/E4i; RE1-silencing transcription factor variant E1b/E2/E3/N3c/E4; RE1-silencing transcription factor variant E1b/E2a/E2k; RE1-silencing transcription factor variant E1b/E2c/E2j/E3/E4; RE1-silencing transcription factor variant E1b/E2e/E4h; RE1-silencing transcription factor variant E1c/E2/E3/E5; RE1-silencing transcription factor variant E1c/E2a/E2k; RE1-silencing transcription factor variant E1c/E2g/E3/E4; repressor binding to the X2 box; Rest; REST4; WT6; X2 box repressor; XBR; zinc finger transcription factor REST protein | |
| Mouse | |
| Protein A | |
| RUO | |
| 5978 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.