Learn More
Cytiva RESOURCE™ Columns for HIC
Excellent for polishing in a purification process
Brand: Cytiva 17-1185-01
Code : GE
Additional Details : CAS Number : 64-17-5 Weight : 0.07600kg
Description
- Prepacked columns for purification of proteins and peptides with hydrophobic interaction chromatography (HIC)
- High performance at low back pressure, even with high sample loads of up to 25mg of protein
- Ligands coupled to 15μm monodispersed rigid polystyrene/divinylbenzene matrix
- Designed for low- to medium- pressure systems ranging from peristaltic pump to AKTA design and FPLC Systems
- RESOURCE PHE will have strongest hydrophobicity followed by RESOURCE ISO and RESOURCE ETH, successively
- Material of column: Polyetheretherketone (PEEK)
- Bed dimensions 6.4 x 30mm
- Maximum pressure 15 bar
- Flow rate: 0.8 to 4.8mL/min
- Storage: 4°C to 30°C, 20% Ethanol
- Matrix: Polystyrene/divinylbenzene
- Stable in common buffers, chaotropic agents, detergents, and polar organic solvents
- Average particle size: 15Glc mum
- pH Stability Working Range: 2 to 12
- Flow velocity: 150cm/h - 900cm/h
Specifications
1 x 1 mL | |
10 cm | |
150 to 900 cm/hr | |
3.937 in. | |
Connectors for high- and low-pressure chromatography equipment and instructions | |
PEEK | |
15 to 15 μm | |
Polystyrene/Divinylbenzene |
3.937 in. | |
RESOURCE ISO column | |
1 mL | |
10 cm | |
Isopropyl | |
15 bar | |
2 to 12 | |
4°C to 30°C, 20% Ethanol |