missing translation for 'onlineSavingsMsg'
Learn More

Cytiva RESOURCE™ Columns for HIC

Product Code. p-3676546 Shop All Cytiva Products
Click to view available options
Bed Volume:
1 mL
1 x 1 mL
Ligand Type:
Isopropyl
Phenyl
Product Type:
RESOURCE ISO column
RESOURCE PHE column
Unit Size:
1mL
This item is not returnable. View return policy

Product Code. 10278944

Brand: Cytiva 17118601

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Connectors for high and low pressure chromatography equipment; Instructions.

Excellent for polishing in a purification process

  • Prepacked columns for purification of proteins and peptides with hydrophobic interaction chromatography (HIC)
  • High performance at low back pressure, even with high sample loads of up to 25mg of protein
  • Ligands coupled to 15μm monodispersed rigid polystyrene/divinylbenzene matrix
  • Designed for low- to medium- pressure systems ranging from peristaltic pump to AKTA design and FPLC Systems
  • RESOURCE PHE will have strongest hydrophobicity followed by RESOURCE ISO and RESOURCE ETH, successively
  • Material of column: Polyetheretherketone (PEEK)
  • Bed dimensions 6.4 x 30mm
  • Maximum pressure 15 bar
  • Flow rate: 0.8 to 4.8mL/min
  • Storage: 4°C to 30°C, 20% Ethanol

Media

  • Matrix: Polystyrene/divinylbenzene
  • Stable in common buffers, chaotropic agents, detergents, and polar organic solvents
  • Average particle size: 15Glc mum
  • pH Stability Working Range: 2 to 12
  • Flow velocity: 150cm/h - 900cm/h

Resource HIC Test Kit (Mfr. No. 17-1187-01) includes one each of 1mL Resource ETH, Resource ISO and Resource PHE columns

TRUSTED_SUSTAINABILITY

Specifications

Bed Volume 1 mL
Diameter (English) Inner 3.937 in.
Diameter (Metric) Inner 10 cm
Product Type RESOURCE PHE column
Flow Rate 150 to 900 cm/hr
Quantity 1 mL
Height (English) 3.937 in.
Height (Metric) 10 cm
Includes Connectors for high- and low-pressure chromatography equipment and instructions
Ligand Type Phenyl
Material PEEK
Max. Pressure 15 bar
Particle Size 15 to 15 μm
pH Range 2 to 12
Support Type Polystyrene/Divinylbenzene
Storage Requirements 4°C to 30°C, 20% Ethanol
Show More Show Less
compliance-icons
Product Identifier
  • RESOURCE PHE, 1 ml
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : ethanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.