missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Resorufin

Product Code. 16475836
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16475836 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16475836 Supplier MedChemExpress Supplier No. HY123533
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Resorufin (NSC 12097) is a highly fluorescent pink dye for the detection of ROS/RNS and a second analyte.

Specifications

Content And Storage Powder -20°C 3 years, 4°C 2 years ∣ In solvent -80°C 6 months, -20°C 1 month
Product Type Dye Reagents
Quantity 100 mg
Shipping Condition Room temperature
Solubility DMF : < 1 mg/mL (ultrasonic) (insoluble) ∣ Ethanol : < 1 mg/mL (ultrasonic) (insoluble) ∣ H2O : < 0.1 mg/mL (ultrasonic) (insoluble) ∣ DMSO : < 1 mg/mL (ultrasonic) (insoluble or slightly soluble)
Target Dopamine Receptor
Molecular Weight (g/mol) 213.19 g/mol
CAS 635-78-9
Synonym NSC 12097
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.