missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Resazurin, Sodium Salt

Product Code. 11523677
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11523677 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 11523677 Supplier Invitrogen™ Supplier No. R12204
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Resazurin, Sodium Salt

The non-fluorescent resazurin can be used as a fluorogenic oxidation-reduction indicator in a variety of cells, including bacteria, yeast and eukaryotes by flow cytometry, fluorescence microscopy and high-throughput screening. The red-fluorescent product, resorufin exhibits absorption/emission maxima ∼575/585 nm.

The non-fluorescent resazurin can be used as a fluorogenic oxidation-reduction indicator in a variety of cells, including bacteria, yeast and eukaryotes by flow cytometry, fluorescence microscopy and high-throughput screening. The red-fluorescent product, resorufin exhibits absorption/emission maxima ∼575/585 nm.
TRUSTED_SUSTAINABILITY

Specifications

Color Red
Content And Storage Store at room temperature and protect from light.
Description Resazurin, Sodium Salt
Detection Method Fluorescence
For Use With (Application) Cell Viability
For Use With (Equipment) Microplate Reader
Product Type Resazurin
Dye Type Resazurin
Form Solid
Product Line Invitrogen
Quantity 10 mg
Shipping Condition Room Temperature.
Excitation Wavelength 575 nm
Emission Wavelength 585
Show More Show Less
How do alamarBlue reagent and PrestoBlue reagent differ from resazurin and C12-resazurin?

alamarBlue reagent and PrestoBlue reagent contain resazurin in a proprietary stabilizing formulation that allows for a convenient “mix, incubate, and read” protocol. PrestoBlue reagent is an improvement in the formulation of alamarBlue reagent that allows for much faster staining (typically 10 minutes vs. 1-4 hours to obtain a similar signal and sensitivity). C12-resazurin is a derivative of resazurin that has better cellular retention and thus allows for analysis on a flow cytometer and multiplexing with viability indicators and other biomarkers.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.