missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Resazurin sodium

Product Code. 16441159
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16441159 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16441159 Supplier MedChemExpress Supplier No. HY111391
 Restricted Item
Estimated Shipment: 28-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Resazurin sodium (Diazoresorcinol sodium) is commonly used to measure bacterial and eukaryotic cell viability through its reduction to the fluorescent product resorufin.

Specifications

Content And Storage 4°C, sealed storage, away from moisture ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture)
Quantity 1 g
Shipping Condition Room temperature
Solubility DMSO : 83.33 mg/mL (331.77 mM; Need ultrasonic) ∣ H2O : 5 mg/mL (19.91 mM; ultrasonic and warming and heat to 60°C)
Molecular Weight (g/mol) 251.17 g/mol
CAS 62758-13-8
Synonym Diazoresorcinol sodium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.