Learn More
Brady™ Repositionable Continuous Vinyl Cloth Label Tape for M6/M7 Printers
Once applied, CleanLift™ Series Vinyl Cloth Labels are permanent or repositionable.
Brand:  Brady™ 173913
 View more versions of this product
Product Code. 17831359
  • ONLINE EXCLUSIVE PRICE
    £97.45
    £92.98 / 1 rolls
    Save £4.47
    4% Off
Estimated Shipment: 08-12-2026
Add to basket
Description
Description
  • Adhesive and cloth backing offers great holding power, yet allow for clean removal and repositioning
  • Sticks to any clean, dry surface
  • Specially formulated topcoat for high print quality
  • Semi-gloss finish for crisp, clean look
  • Made of B-498 vinyl material
Specifications
Specifications
Not Autoclavable
Repositionable Rubber
White
Continuous
Single-side Printable
Label
Asset Tracking, Barcode Labeling, Circuit Board Labeling, Equipment Labeling, Electrical Labeling, General Identification, Wire and Cable Labeling
No
Rectangle
-40°C to +80°C
0.185 mm
30 ft.
0.5 in.
M6C-500-498
ASTM B 117, ASTM D 1000, ASTM D 2979, ASTM G 155, UL Recognized
None
Semi-gloss
Vinyl
Thermal Transfer
Non-sterile
M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter)
30 ft.
-40°F to +176°F
0.0073 in.
30 ft. x 0.5 in.
9.14 m
12.7 mm