missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Replacement Parts for MultiSUB Mini Horizontal Gel System
Shop All Fisherbrand Products
Click to view available options
Type:
Lid
Tank with Electrodes
Unit Size:
Each
Description
MultiSUB Horizontal Gel Electrophoresis Systems combine economical buffer and gel consumption without compromising resolution and separation speed.
The MultiSUB Mini is the preferred option for quick sample checks of small to medium volumes, particularly following restriction digestion during cloning. It is an economical choice for separation of up to 64 samples.
- Electrophoresis Tank (Including Electrodes): A raised platform holds the UV sample trays, with side wells to hold buffer solution; connects to an external power source
- Asymmetrical Safety Lid: Notes the correct polarity and orientation of electrodes, with guides that enable the electrode thumb locators to click easily into the correct position
- Tank and lid are designed for use with both 7 x 7cm or 7 x 10cm gel trays
Order Info
Color
Specifications
Specifications
| Color | Blue |
| For Use With (Application) | Nucleic Acid Gel Electrophoresis |
| For Use With (Equipment) | MultiSUB Mini Horizontal Gel Electrophoresis Unit |
| Type | Lid |
| Gel Dimensions | 7 x 7 cm or 7 x 10 cm |
| Material | Molded Plastic |
| No. of Wells | For up to 64 separations |
| Quantity | 1 Each |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction