missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Replacement Parts for MultiSUB Choice Horizontal Gel System

Product Code. 11873413 Shop All Fisherbrand Products
Click to view available options
Type:
Lid
Tank with Electrodes
Unit Size:
Each

Product Code. 11873413

Only null left
Add to basket
Add to basket

Spare safety lid and electrophoresis tank for use with 15 x 7cm, 15 x 10cm or 15 x 15cm gel trays. Fisherbrand™ Replacement Parts for MultiSUB Choice Horizontal Gel System are constructed of high-quality, injection molded plastic for use in nucleic acid gel electrophoresis applications.

MultiSUB Horizontal Gel Electrophoresis Systems combine economical buffer and gel consumption without compromising resolution and separation speed.

The wider format of the MultiSUB Choice enables the rapid separation of up to 210 samples. A comprehensive selection of combs provides full flexibility for preparatory and screening techniques. MS Choice is suitable for cloning and mini-prep analysis, and it is ideal for restriction fragment analysis, sample prep or checking of high numbers of samples.

  • Electrophoresis Tank (Including Electrodes): A raised platform holds the UV sample trays, with side wells to hold buffer solution; connects to an external power source
  • Asymmetrical Safety Lid: Notes the correct polarity and orientation of electrodes, with guides that enable the electrode thumb locators to click easily into the correct position
  • Tank and lid are designed for use with 15 x 7cm, 15 x 10cm or 15 x 15cm gel trays

Specifications

Color Clear
For Use With (Application) Nucleic Acid Gel Electrophoresis
For Use With (Equipment) MultiSUB Choice Horizontal Gel Electrophoresis Unit
Type Tank with Electrodes
Gel Dimensions 15 x 7 cm, 15 x 10 cm or 15 x 15 cm
Material Molded Plastic
No. of Wells For up to 210 separations
Quantity 1 Each
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.