missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Replacement Parts for MultiSUB Choice Horizontal Gel System

Product Code. 11813303 Shop All Fisherbrand Products
Click to view available options
Type:
Lid
Tank with Electrodes
Unit Size:
Each

Product Code. 11813303

Brand: Fisherbrand™ FMS15LID

Only null left
Add to basket
Add to basket

Spare safety lid and electrophoresis tank for use with 15 x 7cm, 15 x 10cm or 15 x 15cm gel trays. Fisherbrand™ Replacement Parts for MultiSUB Choice Horizontal Gel System are constructed of high-quality, injection molded plastic for use in nucleic acid gel electrophoresis applications.

MultiSUB Horizontal Gel Electrophoresis Systems combine economical buffer and gel consumption without compromising resolution and separation speed.

The wider format of the MultiSUB Choice enables the rapid separation of up to 210 samples. A comprehensive selection of combs provides full flexibility for preparatory and screening techniques. MS Choice is suitable for cloning and mini-prep analysis, and it is ideal for restriction fragment analysis, sample prep or checking of high numbers of samples.

  • Electrophoresis Tank (Including Electrodes): A raised platform holds the UV sample trays, with side wells to hold buffer solution; connects to an external power source
  • Asymmetrical Safety Lid: Notes the correct polarity and orientation of electrodes, with guides that enable the electrode thumb locators to click easily into the correct position
  • Tank and lid are designed for use with 15 x 7cm, 15 x 10cm or 15 x 15cm gel trays

Specifications

Color Blue
For Use With (Application) Nucleic Acid Gel Electrophoresis
For Use With (Equipment) MultiSUB Choice Horizontal Gel Electrophoresis Unit
Type Lid
Gel Dimensions 15 x 7 cm, 15 x 10 cm or 15 x 15 cm
Material Molded Plastic
No. of Wells For up to 210 separations
Quantity 1 Each
Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato