Learn More
Mettler Toledo™ Replacement Parts for C20 Karl Fischer™ Autotitrator
Accessories For use with Mettler Toledo C20 Karl Fischer™ Autotitrator
Brand:  Mettler Toledo™ 51108751
Code : 04
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 12655815
  • £2980.93 / Each
Estimated Shipment: 06-06-2024
to see stock.
Add to basket
Description
Description
Coulometric Cells
  • Coulometer cells with or without diaphragm
  • Cell without diaphragm is almost maintenance-free:, innovative design lets you use this cell for determination of water in oils.
  • Cell with a diaphragm is recommended for applications such as the determination of water in substances containing ketones, also recommended for best possible accuracy
Coulometric Reagent Exchange Set
Allows emptying and re-filling the coulometric cell, reducing the risk of the user coming into contact with reagents
Specifications
Specifications
Coulometric Cell with Diaphragm
Ketones, aldehydes, very low water values or where best possible accuracy is needed
Glass