missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ Replacement Parts for C20 Karl Fischer™ Autotitrator

Product Code. p-4684528 Shop All Mettler Toledo Products
Click to view available options
Type:
Coulometric Cell with Diaphragm
Coulometric Cell without Diaphragm
Drying Tube
Septa
Solvent Drain/Delivery Tube
Unit Size:
Each
Pack of 12
This item is not returnable. View return policy

Product Code. 11390532

Brand: Mettler Toledo™ 51108753

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Accessories For use with Mettler Toledo C20 Karl Fischer™ Autotitrator

Coulometric Cells

  • Coulometer cells with or without diaphragm
  • Cell without diaphragm is almost maintenance-free:, innovative design lets you use this cell for determination of water in oils.
  • Cell with a diaphragm is recommended for applications such as the determination of water in substances containing ketones, also recommended for best possible accuracy
Coulometric Reagent Exchange Set
Allows emptying and re-filling the coulometric cell, reducing the risk of the user coming into contact with reagents
TRUSTED_SUSTAINABILITY

Specifications

Type Coulometric Cell without Diaphragm
For Use With (Application) General coulometric KF titration
Material Glass
For Use With (Equipment) C20 Coulometric Titrator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.