missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trajan™ Shimadzu Splitless Straight-Through Liner
The unique deactivation process of SGE inlet liners provides excellent analysis for a wide range of compounds
Brand: Trajan™ 092086101
Code : 13
Additional Details : Weight : 0.10000kg
Description
- Each batch of liners tested for activity
- Manufactured from borosilicate glass
- Color coded by geometry for easy identification
- For use with Shimazdu instruments
Specifications
GC Inlet Liner, Splitless, Straight-Through | |
95 mm | |
5 mm |
1 Ea. | |
2.6 mm |