Learn More
Labconco™ Glass Manifold
Make exhaustion of fumes from Kjeldahl digestions more convenient by adding a Labconco Glass Manifold to your system.
Brand: Labconco™ 6001100
Code : NEW
Additional Details : Weight : 2.00000kg
Description
- Six-port glass manifold exhausts fumes from Kjeldahl digestions
- Serrated connector for water aspirator
Specifications
Manifold | |
Glass |