missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Replacement Items for Freezer/Mill™ Grinding Vial Sets

Product Code. 12598686 Shop All SPEX Certiprep Products
Click to view available options
Product Type:
Center Cylinder
End Plug
Replacement Impactor
Material:
Polycarbonate
Stainless Steel (440 C)
For Use With (Equipment):
6801 Large Grinding Vial Set and 6871 Large Cr-Free Grinding Vial Set
6881 Vial Set
Spex SamplePrep Freezer/Mill Cryogenic Grinder
Unit Size:
Each
Pack of 2
Pack of 20
Pack of 4
This item is not returnable. View return policy

Product Code. 12598686

Brand: Cole-Parmer™ 6751P

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Keep spare items to ensure there is no down time.

  • Suitable for use with the Cole-Parmer™ Freezer/Mill™ grinding vial sets
  • Optimize performance of your Cole-Parmer™ Freezer/Mill™ grinding vial sets with a wide range of grinding ball options suitable for your application.

Specifications

Product Type Replacement Impactor
Autoclavable Autoclavable
For Use With (Equipment) Spex SamplePrep Freezer/Mill Cryogenic Grinder
Material Stainless Steel (440 C)
Model Cole-Parmer Freezer/Mill
Packaging Quantity 1 Ea.
Sample Volume (English) 0.01 to 0.13 oz.
Sample Volume (Metric) 0.5 to 4 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.