Learn More
Idex Replacement Ferrules for SealTight™ Fittings
Slotted end on ferrule creates second compression zone for a tight seal
£7.85 - £54.91
Specifications
Color | Black |
---|---|
Description | 10-32 or M6 coned, for 1/16 in. OD tubing |
Material | PEEK |
Type | Ferrule |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11593970
|
Idex
F-192 |
1/Ea |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11588670
|
Idex
F-192X |
10/pk |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Black | |
10-32 or M6 coned, for 1/16 in. OD tubing | |
PEEK | |
Ferrule |