Learn More
Idex Replacement Ferrules for SealTight™ Fittings
Slotted end on ferrule creates second compression zone for a tight seal
Brand:  Idex F-192
Code : 16
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 11593970
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Specifications
10-32 or M6 coned, for 1/16 in. OD tubing
PEEK
1/Ea
Black
Ferrule