missing translation for 'onlineSavingsMsg'
Learn More

Trajan™ Replaceable Plunger in Needle Kit

SGE NanoVolume syringes are designed to inject down to 50nL with high precision and accuracy

Brand:  Trajan™ 033708

Code : 13

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10442731

  • £47.60 / Each
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • High precision and accuracy for small volume injections
  • Replacement plunger/needle kits
  • Kits are matched. Needles and plungers must be replaced as a set
Specifications

Specifications

Stainless Steel (Tip)
0.47 mm
Replacement Needle and Plunger: NP0.5B-HP-0.47
42 mm
Syringes (0.5μL)
26 G
0.47 mm
0.5 μL
Cone
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers