Learn More
BRAND™ Repair Set for Transferpettor™
Includes everything necessary to make small repairs to the BRAND™ Transferpettor™
Brand:  BRAND™ 701967
Additional Details : Weight : 0.02720kg
 View more versions of this product
Product Code. 15633987
  • £40.50 / Each
Estimated Shipment: 24-05-2024
to see stock.
Add to basket
Description
Description
  • Includes 1 allen key, 1 piston rod with fitted PTFE seal, 1 calibrating gauge, 1 screwdriver, 3 clamping discs, 1 fixing screw, 3 PTFE seals
  • Mounting block included for capacities of 20μL and up
Specifications
Specifications
DE-M marked
Transferpettor™
1 Each
50 μL
Green mounting block