Learn More
BRAND™ Repair Set for Transferpettor™
Includes everything necessary to make small repairs to the BRAND™ Transferpettor™
£36.10 - £68.45
Specifications
Certifications/Compliance | DE-M marked |
---|---|
For Use With (Equipment) | Transferpettor™ |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
10232511
|
BRAND™
701968 |
100 and 200 μL |
£40.50
Each |
In Stock
Log in to see stock. |
|||||
10191771
|
BRAND™
701965 |
10 μL |
£36.10
Each |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
15633987
|
BRAND™
701967 |
50 μL |
£40.50
Each |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
10726901
|
BRAND™
701966 |
20 and 25 μL |
£68.45
Each |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
10538781
|
BRAND™
701964 |
1, 2, 5 μL |
£36.45
Each |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
10130571
|
BRAND™
701967 |
50 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Includes 1 allen key, 1 piston rod with fitted PTFE seal, 1 calibrating gauge, 1 screwdriver, 3 clamping discs, 1 fixing screw, 3 PTFE seals
- Mounting block included for capacities of 20μL and up
Specifications
DE-M marked | |
Transferpettor™ |