Learn More
BRAND™ Repair Set for Transferpettor™
Includes everything necessary to make small repairs to the BRAND™ Transferpettor™
Brand: BRAND™ 701964
Code : 03
Additional Details : Weight : 0.01000kg
Description
- Includes 1 allen key, 1 piston rod with fitted PTFE seal, 1 calibrating gauge, 1 screwdriver, 3 clamping discs, 1 fixing screw, 3 PTFE seals
- Mounting block included for capacities of 20μL and up
Specifications
DE-M marked | |
Transferpettor™ | |
1Each |
1, 2, 5 μL | |
No mounting block | |
1 Each |