Learn More
Cole-Parmer™ Repair Kit for Vacuum/Pressure Pump
Product Code. 11725888 Shop All Cole-Parmer Products
Change view
Click to view available options
Material:
Zinc Alloy
Unit Size:
Each
Product Code. Material unitSize
11725888 Zinc Alloy Each
1 options
Product Code. 11725888
Brand: Cole-Parmer™ 7930163
Only null left
Add to basket
Add to basket
Use the Repair Kit for Vacuum/Pressure Pump to produce up to 25 in. Hg vacuum or test for pressure leaks.
  • Pumps are sealed and self-lubricated for a long, maintenance-free life.
  • Vacuum gauge is graduated in both centimeters and inches of mercury.
  • Built-in trigger enables you to release vacuum without disconnecting the pump.
  • Zinc alloy is perfect for rugged applications; PVC is lightweight and easy to handle.
Specifications
For Use With (Application) Repairing Vacuum/Pressure Pumps
For Use With Vacuum/Pressure Pump 79301-11
Material Zinc Alloy
Quantity 1 Ea.
Type Repair Kit