missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Remel™ Tryptic Soy Broth

Product Code. 17719448
Change view
Click to view available options
Quantity:
10 x 100 mL
Format:
BB Septum - 100mL
Packaging Type:
10/Pk.
Unit Size:
Pack of 10
Product Code. Quantity Format Packaging Type unitSize
17719448 10 x 100 mL BB Septum - 100mL 10/Pk. Pack of 10
1 options
This item is not returnable. View return policy

Product Code. 17719448

Brand: Thermo Scientific™ R112745

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The Thermo Scientific™ Remel Tryptic Soy Broth (TSB) is a general purpose enrichment broth for cultivation and isolation of microorganisms.

Thermo Scientific™ Remel Tryptic Soy Broth (TSB) is a general purpose enrichment broth for cultivation and isolation of microorganisms.

  • Part numbers beginning with R11 are for sterility testing according to USP (United States Pharmacopeia) and for use as general purpose enrichment broth
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: AOAC, BAM, COMPF, SMWW, IDF, SMD, USDA, USP.
Description Tryptic Soy Broth
Format BB Septum - 100mL
Product Type Microbiology Media
Packaging Type 10/Pk.
Quantity 10 x 100 mL

REF numbers beginning with R11 are not intended for IVD use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.