missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Settling Plate Tryptic Soy Agar

Product Code. 17425539
Click to view available options
Quantity:
10/Pk.
Format:
100mm Plate, Double
85mm Plate, Double
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 17425539

Brand: Thermo Scientific™ R111879

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Isolate and cultivate wide variety of organisms like bacteria, yeast and fungi using Settling Plate Tryptic Soy Agar.

Isolate and cultivate wide variety of organisms such as bacteria, yeast and fungi using Thermo Scientific™ Remel™ Settling Plate Tryptic Soy Agar. This medium is listed in Standard Methods for the Examination of Water and Wastewater and the Compendium of Methods for the Microbiological Examination of Foods1,2. This medium is recommended for use in Official Methods of Analysis of AOAC International, the International Dairy Federation (IDF), and the United States Department of Agriculture (USDA)3,4. Tryptic Soy Agar slants may be used to cultivate, store, and ship bacterial cultures.

Tryptic Soy Agar is prepared following the standard formula for Soybean-Casein Digest Agar Medium in the United States of Pharmacopeia (USP)5. Tryptic Soy Agar many be supplemented with blood to facilitate the growth of more fastidious bacteria or antimicrobial agents to permit the selection of various microbial groups from mixed flora.

  • Recommended for use in Official Methods of Analysis of AOAC International, IDF, and USDA3,4
  • Ready to use – Convenience of prepared medium

This medium contains casein and soy peptones that provide nitrogen, amino acids, and peptides necessary for bacterial growth. Dextrose is a ready source of energy. Sodium chloride supplies essential electrolytes and maintains osmotic equilibrium.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Settling Plate Tryptic Soy Agar
Format 100mm Plate, Double
Product Type Agar
Quantity 10/Pk.

Not intended for IVD use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.