Learn More
Thermo Scientific™ Remel™ Contact Plate Sterile Tryptic Soy Agar w/Lecithin, Polysorbate 80 (Irradiated)
Product Code. 17295954
Change view
Click to view available options
Quantity:
10/Pk.
Unit Size:
Pack of 10
Product Code. Quantity unitSize
17295954 10/Pk. Pack of 10
1 options
Product Code. 17295954 Supplier Thermo Scientific™ Supplier No. R111800
Thermo Scientific™ Remel Contact Plate Sterile Tryptic Soy Agar with Lecithin, Polysorbate 80 (Irradiated) is for the cultivation of bacteria, yeast and fungi.
  • Double-bagged plus a third bag is enclosed for customer use (Double + Bag)
  • Products are manufactured in a clean room and validated sterile by gamma radiation
  • Made using GripFit™ technology, which ensures the lid remains in place during handling and yet can be easily removed during environmental monitoring
TRUSTED_SUSTAINABILITY
Specifications
Description Contact Plates; Sterile; Tryptic Soy Agar w/Lecithin; Gamma irradiated
Format Contact - Double Bag
Product Type Agar
Quantity 10/Pk.
Not intended for IVD use.