missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Remel™ Butterfield's Phosphate Buffer

Product Code. p-4522922
Click to view available options
Quantity:
10 x 100 mL
Format:
PB -100mL
Packaging Type:
10/Pk.
Unit Size:
1 L
This item is not returnable. View return policy

Product Code. 30238167

Brand: Thermo Scientific™ R112519

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific™ Remel Butterfield's Phosphate Buffer (pH 7.2) is used as a diluent in microbial limit testing of pharmaceutical products and food testing.

Thermo Scientific™ Remel Butterfield's Phosphate Buffer (pH 7.2) is used as a diluent in microbial limit testing of pharmaceutical products and food testing.

Not intended for IVD use.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: BAM, USDA, USP.
Description Phosphate Buffer, Butterfield's
Format PB -100mL
Product Type Buffer Solution
Packaging Type 10/Pk.
Quantity 10 x 100 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.