missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Remel™ Butterfield's Phosphate Buffer

Product Code. 30238167
Change view
Click to view available options
Quantity:
10 x 100 mL
Format:
PB -100mL
Packaging Type:
10/Pk.
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Format Packaging Type unitSize
30238167 10 x 100 mL PB -100mL 10/Pk. 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30238167 Supplier Thermo Scientific™ Supplier No. R112519

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific™ Remel Butterfield's Phosphate Buffer (pH 7.2) is used as a diluent in microbial limit testing of pharmaceutical products and food testing.

Thermo Scientific™ Remel Butterfield's Phosphate Buffer (pH 7.2) is used as a diluent in microbial limit testing of pharmaceutical products and food testing.

Not intended for IVD use.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: BAM, USDA, USP.
Description Phosphate Buffer, Butterfield's
Format PB -100mL
Product Type Buffer Solution
Packaging Type 10/Pk.
Quantity 10 x 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.