missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Remel™ BHI Broth (Brain Heart Infusion Broth)

Product Code. 30238151
Change view
Click to view available options
Quantity:
20 x 4 mL
20 x 5 mL
100 x 4 mL
100 x 5 mL
Format:
15mm x 103mm Tube
Unit Size:
Pack of 100
Pack of 20
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Format unitSize
30238151 100 x 5 mL 15mm x 103mm Tube -
17742998 100 x 4 mL 15mm x 103mm Tube Pack of 100
17752998 20 x 4 mL 15mm x 103mm Tube Pack of 20
17762998 100 x 5 mL 15mm x 103mm Tube Pack of 100
17772998 20 x 5 mL 15mm x 103mm Tube Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 30238151 Supplier Thermo Scientific™ Supplier No. R060270

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific™ Remel BHI Broth (Brain Heart Infusion Broth) is for the cultivation of microorganisms.

Thermo Scientific™ Remel BHI Broth (Brain Heart Infusion Broth) is for the cultivation of microorganisms.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: BAM, SMD, SMWW, USDA.
Description BHI Broth
Format 15mm x 103mm Tube
Product Type Microbiology Media
Quantity 100 x 5 mL

REF R14101 for use with MicroScan™ System. REF R112022 not intended for IVD use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.