missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Remel™ BHI Broth (Brain Heart Infusion Broth)
Click to view available options
Quantity:
20 x 4 mL
20 x 5 mL
100 x 4 mL
100 x 5 mL
Format:
15mm x 103mm Tube
Unit Size:
Pack of 100
Pack of 20
Description
Thermo Scientific™ Remel BHI Broth (Brain Heart Infusion Broth) is for the cultivation of microorganisms.
Specifications
Specifications
| Certifications/Compliance | Industrial Reference: BAM, SMD, SMWW, USDA. |
| Description | BHI Broth |
| Format | 15mm x 103mm Tube |
| Product Type | Microbiology Media |
| Quantity | 20 x 5 mL |
REF R14101 for use with MicroScan™ System. REF R112022 not intended for IVD use.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction