missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Relaxin 3 Monoclonal Antibody (D10)

Mouse Monoclonal Antibody
£178.00 - £486.00
Specifications
| Antigen | Relaxin 3 |
|---|---|
| Clone | D10 |
| Concentration | 1 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17768313
![]() |
Invitrogen™
MA541667 |
200 μL |
£486.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17654066
|
Invitrogen™
MA543609 |
20 μL |
£178.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Relaxin 3 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
Relaxin 3 is part of the Relaxin-like peptide family that plays an important neuropeptide signaling role. Relaxin 3 is a ligand for two structurally related G protein-coupled receptors, human Relaxin receptor 3 and Relaxin receptor 4. The greatest levels of Relaxin 3 expression occur in the brain, specifically in the pons/medulla, with highest levels in the pars ventromedialis of the dorsal tegmental nucleus and with lower levels in the hippocampus and olfactory regions. Significant expression is also observed in the spleen, thymus, lung and ovary. Relaxin 3 inhibits Forskolin-stimulated cAMP accumulation in a dose-dependent manner in cells that express Relaxin receptor 3. Acute and repeated administration of Relaxin 3 causes a decrease in levels of the plasma thyroid stimulating hormone, suggesting that Relaxin 3 may play a role in long-term control of food intake.Specifications
| Relaxin 3 | |
| 1 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q8BFS3, Q8WXF3 | |
| 117579, 266997 | |
| Recombinant protein Relaxin 3. The antigen corresponds to amino acid range 1-140 of the target protein. | |
| Antibody |
| D10 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Monoclonal | |
| Liquid | |
| IgG2b κ | |
| Human, Rat | |
| RLN3 | |
| H3; insl7; insulin-like 7; insulin-like peptide 7; Insulin-like peptide INSL7; M3; PRO20213; Prorelaxin H3; prorelaxin M3; relaxin 3; relaxin-3; Relaxin-3 A chain; Relaxin-3 B chain; RLN3; RXN3; UNQ6188; UNQ6188/PRO20213; ZINS4 | |
| RLN3 | |
| Primary | |
| Protein A/G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title