Learn More
Abnova™ RELA/CEN11p FISH Probe
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
Brand: Abnova™ FG0221.200uL
Additional Details : Weight : 0.02000kg
Specifications
Store at 4°C in the dark | |
Fluorescent In Situ Hybridization (Cell) | |
MGC131774,NFKB3,p65 | |
5970 | |
200 μL |
Tube | |
v-rel reticuloendotheliosis viral oncogene homolog A (avian) | |
RELA | |
DNA/RNA | |
Liquid |