missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ RELA/CEN11p FISH Probe
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
£1235.00 - £2030.00
Specifications
Content And Storage | Store at 4°C in the dark |
---|---|
Format | Tube |
For Use With (Application) | Fluorescent In Situ Hybridization (Cell) |
Gene | v-rel reticuloendotheliosis viral oncogene homolog A (avian) |
Gene Alias | MGC131774,NFKB3,p65 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16003365
|
Abnova™
FG0221.200UL |
200 μL |
£2030.00
200µL |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16093355
|
Abnova™
FG0221.100UL |
100 μL |
£1235.00
100µL |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Store at 4°C in the dark | |
Fluorescent In Situ Hybridization (Cell) | |
MGC131774,NFKB3,p65 | |
5970 | |
Liquid |
Tube | |
v-rel reticuloendotheliosis viral oncogene homolog A (avian) | |
RELA | |
DNA/RNA |