Learn More
Gilac™ Reinforced Stacking and Nesting Container
Description
- Reinforced containers for intensive use and extreme handling conditions.
- Stackable and nestable by a quarter turn.
- Highly durable.
- Suitable for a number of different uses in the food processing, fishmonger, butchery, charcuterie, dairy and waste collection industries.
Specifications
Specifications
| Type | Storage Tote |
| Material | HDPE |
| Color | White |
| Shape | Rectangular |
| Capacity (English) | 15.9 gal. |
| Capacity (Metric) | 60 L |