missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Regulatory protein GAL4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£153.00 - £378.00
Specifications
| Antigen | Regulatory protein GAL4 |
|---|---|
| Concentration | 1.0 mg/ml |
| Dilution | Western Blot 1 - 2 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18288918
|
Novus Biologicals
NBP2-76808SS |
0.025 mg |
£153.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18283269
|
Novus Biologicals
NBP2-76808 |
0.1 mg |
£378.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Regulatory protein GAL4 Polyclonal specifically detects Regulatory protein GAL4 in Yeast samples. It is validated for Western Blot.Specifications
| Regulatory protein GAL4 | |
| Western Blot 1 - 2 ug/ml | |
| Polyclonal | |
| Rabbit | |
| PBS with 0.02% sodium azide | |
| IgG | |
| Affinity Purified |
| 1.0 mg/ml | |
| Western Blot | |
| Unconjugated | |
| Yeast | |
| Partial recombinant protein made to a N terminal portion of the yeast GAL4 protein (amino acids 1-147) [UniProt P04386] | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title